Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Aminohexyl)phosphonic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
63207-60-3
C6H16NO3P
181.17
MFCD00799469
ODMTWOKJHVPSLP-ZCFIWIBFSA-N
1r---1-aminohexyl phosphonic acid,phosphonic acid,p-1-aminohexyl,r-1-aminohexyl phosphonic acid,1r-1-aminohexyl phosphonic acid,1r-1-aminohexylphosphonic acid
7021160
[(1R)-1-aminohexyl]phosphonic acid
CCCCCC(N)P(=O)(O)O
63207-60-3
C6H16NO3P
181.17
MFCD00799469
ODMTWOKJHVPSLP-ZCFIWIBFSA-N
1r---1-aminohexyl phosphonic acid,phosphonic acid,p-1-aminohexyl,r-1-aminohexyl phosphonic acid,1r-1-aminohexyl phosphonic acid,1r-1-aminohexylphosphonic acid
7021160
[(1R)-1-aminohexyl]phosphonic acid
CCCCCC(N)P(=O)(O)O
Infrared Spectrum Authentic
Color White
Melting Point 265.0°C to 268.0°C
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)4CH(NH2)P(O)(OH)2
Solubility Information Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
Formula Weight 181.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (1-Aminohexyl)phosphonic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.