Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminohomopiperidine 97.0+%, TCI America™

Click to view available options
Quantity:
5g
5906-35-4
C6H14N2
114.192
MFCD00003260
UGBKOURNNQREPE-UHFFFAOYSA-N
1-aminohomopiperidine,1h-azepin-1-amine, hexahydro,n-aminohomopiperidine,n-aminohexamethylenimine,n-aminohexamethyleneimine,aminohexamethyleneimine,1,1-hexamethylenehydrazine,1-aminoperhydroazepine,1-aminohexahydroazepine,1-aminohexamethyleneimine
22198
azepan-1-amine
C1CCCN(CC1)N
5906-35-4
C6H14N2
114.192
MFCD00003260
UGBKOURNNQREPE-UHFFFAOYSA-N
1-aminohomopiperidine,1h-azepin-1-amine, hexahydro,n-aminohomopiperidine,n-aminohexamethylenimine,n-aminohexamethyleneimine,aminohexamethyleneimine,1,1-hexamethylenehydrazine,1-aminoperhydroazepine,1-aminohexahydroazepine,1-aminohexamethyleneimine
22198
azepan-1-amine
C1CCCN(CC1)N
Color Colorless
Quantity 5g
Boiling Point 100°C
UN Number 2733
Formula Weight 114.19
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 1-Aminohomopiperidine

RTECSNumber : CM3165000

TSCA : No