Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminohydantoin hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2827-56-7
C3H5N3O2·ClH
151.55
WEOHANUVLKERQI-UHFFFAOYSA-N
1-aminohydantoin hydrochloride,1-aminoimidazolidine-2,4-dione hydrochloride,1-aminohydantoin hcl,1-amino hydantoin hydrochloride,2,4-imidazolidinedione, 1-amino-, monohydrochloride,acmc-209h1m,1-aminoimidazolidine-2,4-dione hcl,1-aminoimidazoline-2,4-dione hydrochloride
12472963
1-aminoimidazolidine-2,4-dione;hydrochloride
C1C(=O)NC(=O)N1N.Cl
2827-56-7
C3H5N3O2·ClH
151.55
WEOHANUVLKERQI-UHFFFAOYSA-N
1-aminohydantoin hydrochloride,1-aminoimidazolidine-2,4-dione hydrochloride,1-aminohydantoin hcl,1-amino hydantoin hydrochloride,2,4-imidazolidinedione, 1-amino-, monohydrochloride,acmc-209h1m,1-aminoimidazolidine-2,4-dione hcl,1-aminoimidazoline-2,4-dione hydrochloride
12472963
1-aminoimidazolidine-2,4-dione;hydrochloride
C1C(=O)NC(=O)N1N.Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 201.0°C to 205.0°C
Quantity 25g
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5% (Argentometry)
Formula Weight 151.55
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Aminohydantoin hydrochloride
Show More Show Less