Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminomethyl-1-cyclohexanol hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
19968-85-5
C7H15NO·HCl
165.67
MFCD00003859
DXIAEURKHWGAFH-UHFFFAOYSA-N
1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride
3084332
1-(aminomethyl)cyclohexan-1-ol;hydrochloride
C1CCC(CC1)(CN)O.Cl
19968-85-5
C7H15NO·HCl
165.67
MFCD00003859
DXIAEURKHWGAFH-UHFFFAOYSA-N
1-aminomethyl-1-cyclohexanol hydrochloride, 1-aminomethyl cyclohexanol hydrochloride, 1-aminomethylcyclohexan-1-ol hydrochloride, 1-aminomethyl-cyclohexanol hydrochloride, 1-aminomethyl cyclohexan-1-ol hydrochloride, 1-aminomethylcyclohexan-1-ol hcl, acmc-20aoy8, 1-aminomethyl-1-cyclohexanol hcl, 1-aminomethylcyclohexanol hydrochloride, 1-aminomethyl-cyclohexanol hydrogen chloride
3084332
1-(aminomethyl)cyclohexan-1-ol;hydrochloride
C1CCC(CC1)(CN)O.Cl
Quantity 5g
Color Beige-Gray to White
Melting Point 217°C to 219°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula H2NCH2C6H10OH·HCl
Formula Weight 165.67
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1-Aminomethyl-1-cyclohexanol hydrochloride, 98%
Show More Show Less

EINECSNumber : 243-444-2

TSCA : TSCA