Alfa Aesar™ 1-Aminopyrene, 97%

Manufacturer: Alfa Aesar™ 4396802

Quantity 0.5g

 View more versions of this product

Catalog No. AA4396802

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1606-67-3
    Molecular Formula C16H11N
    Formula Weight 217.27
    MDL Number MFCD00004140
    Chemical Name or Material 1-Aminopyrene

    Specifications

    Formula Weight 217.27
    Health Hazard 3 GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1606-67-3
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    Quantity 0.5g
    Molecular Formula C16H11N
    Melting Point 116° to 119°C
    MDL Number MFCD00004140
    Health Hazard 2 GHS H Statement
    H341-H302
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Chemical Name or Material 1-Aminopyrene
    Assay 97%

    GHS H Statement
    H341-H302
    Suspected of causing genetic defects.
    Harmful if swallowed.

    GHS P Statement
    P281-P264-P301+P312-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Wash thoroughly after handling.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit