Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Aza-15-crown-5, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.
Solubility
Soluble in water and methanol.
Notes
Air sensitive. Store away from oxidizing agents.
1-Aza-15-crown-5 is used as building block for the synthesis of bis-crown ethers, lariat ethers and other monofuctionalized macrocycles. It is also used as pharmaceutical intermediate.
Solubility
Soluble in water and methanol.
Notes
Air sensitive. Store away from oxidizing agents.
Specifications
Quantity | 5 g |
Beilstein | 971959 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 219.28 |
Percent Purity | 97% |
Chemical Name or Material | 1-Aza-15-crown-5 |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 266-523-3 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction