Alfa Aesar™ 1-Azaxanthone, 97%

Manufacturer: Alfa Aesar™ A1204514

Quantity 25g

 View more versions of this product

Catalog No. AAA1204514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6537-46-8
    Molecular Formula C12H7NO2
    Formula Weight 197.2
    MDL Number MFCD00052253
    Beilstein 8657
    Chemical Name or Material 1-Azaxanthone

    Specifications

    Formula Weight 197.2
    MDL Number MFCD00052253
    Molecular Formula C12H7NO2
    CAS 6537-46-8
    Beilstein 8657
    Melting Point 180° to 182°C
    Quantity 25g
    Chemical Name or Material 1-Azaxanthone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit