Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzoylnaphthalene, 97%

Click to view available options
Quantity:
1g
5g
25g
642-29-5
C17H12O
232.282
MFCD00046431
CXAYOCVHDCXPAI-UHFFFAOYSA-N
1-benzoylnaphthalene,1-naphthyl phenyl ketone,benzoylnaphthalene,naphthalen-1-yl phenyl methanone,naphthyl phenyl ketone,1-naphthyl phenyl methanone,alpha-benzoylnaphthalene,methanone, 1-naphthalenylphenyl,naphthylphenylketone
69503
naphthalen-1-yl(phenyl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=CC3=CC=CC=C32
642-29-5
C17H12O
232.282
MFCD00046431
CXAYOCVHDCXPAI-UHFFFAOYSA-N
1-benzoylnaphthalene,1-naphthyl phenyl ketone,benzoylnaphthalene,naphthalen-1-yl phenyl methanone,naphthyl phenyl ketone,1-naphthyl phenyl methanone,alpha-benzoylnaphthalene,methanone, 1-naphthalenylphenyl,naphthylphenylketone
69503
naphthalen-1-yl(phenyl)methanone
C1=CC=C(C=C1)C(=O)C2=CC=CC3=CC=CC=C32
Quantity 1g
Melting Point 75°C to 76°C
Formula Weight 232.28
Percent Purity 97%
Chemical Name or Material 1-Benzoylnaphthalene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning