Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzoylpiperazine +98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13754-38-6
C11H14N2O
190.246
MFCD00810192
VUNXBQRNMNVUMV-UHFFFAOYSA-N
1-benzoylpiperazine,n-benzoylpiperazine,phenyl-piperazin-1-yl-methanone,1-benzoyl piperazine,1-benzoyl-piperazine,phenyl piperazin-1-yl methanone,piperazine, 1-benzoyl,phenyl piperazinyl ketone,phenyl piperazin-4-ium-1-yl methanone,1-phenylcarbonyl piperazine
762654
phenyl(piperazin-1-yl)methanone
C1CN(CCN1)C(=O)C2=CC=CC=C2
13754-38-6
C11H14N2O
190.246
MFCD00810192
VUNXBQRNMNVUMV-UHFFFAOYSA-N
1-benzoylpiperazine,n-benzoylpiperazine,phenyl-piperazin-1-yl-methanone,1-benzoyl piperazine,1-benzoyl-piperazine,phenyl piperazin-1-yl methanone,piperazine, 1-benzoyl,phenyl piperazinyl ketone,phenyl piperazin-4-ium-1-yl methanone,1-phenylcarbonyl piperazine
762654
phenyl(piperazin-1-yl)methanone
C1CN(CCN1)C(=O)C2=CC=CC=C2
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Yellow
Melting Point 62°C to 66°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 190.25
Physical Form Crystalline Powder
Percent Purity ≥98.0%
Chemical Name or Material 1-Benzoylpiperazine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger