Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Benzyl-2-pyrrolidinone, 97%
Click to view available options
Quantity:
25g
100g
Description
Applications
1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
Flash Point | >110°C (230°F) |
Refractive Index | 1.552 |
Density | 1.095 |
Quantity | 100g |
Boiling Point | 76°C to 78°C (0.1mmHg) |
Beilstein | 132168 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 175.23 |
Percent Purity | 97% |
Chemical Name or Material | 1-Benzyl-2-pyrrolidinone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H320-H335
EINECSNumber : 226-131-5
TSCA : No
Recommended Storage : Ambient temperatures