Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-piperidinol, 97%, Maybridge

Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
105973-51-1
C12H18ClNO
227.732
MFCD00044200
PHJDPNCYJSWYGY-UHFFFAOYSA-N
1-benzyl-3-piperidinol hydrochloride,1-benzylpiperidin-3-ol hydrochloride,1-benzyl-3-piperidinol hcl,1-benzyl-3-piperidinolhydrochloride,1-benzyl-3-hydroxypiperidine hydrochloride,acmc-1ca0o,1-benzylpiperidin-3-ol, chloride,1-benzyl3-hydroxypiperidine hydrochloride,1-phenylmethyl-3-piperidinol hydrochloride,1-phenylmethyl piperidin-3-ol hydrochloride
16218306
1-benzylpiperidin-3-ol;hydrochloride
C1CC(CN(C1)CC2=CC=CC=C2)O.Cl
105973-51-1
C12H18ClNO
227.732
MFCD00044200
PHJDPNCYJSWYGY-UHFFFAOYSA-N
1-benzyl-3-piperidinol hydrochloride,1-benzylpiperidin-3-ol hydrochloride,1-benzyl-3-piperidinol hcl,1-benzyl-3-piperidinolhydrochloride,1-benzyl-3-hydroxypiperidine hydrochloride,acmc-1ca0o,1-benzylpiperidin-3-ol, chloride,1-benzyl3-hydroxypiperidine hydrochloride,1-phenylmethyl-3-piperidinol hydrochloride,1-phenylmethyl piperidin-3-ol hydrochloride
16218306
1-benzylpiperidin-3-ol;hydrochloride
C1CC(CN(C1)CC2=CC=CC=C2)O.Cl
Infrared Spectrum Authentic
Packaging Amber Glass Bottle
Quantity 10g
Color White
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 227.73
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1-Benzyl-3-piperidinol hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.