Alfa Aesar™ 1-Benzyl-4-chlorophthalazine, 99%

Manufacturer: Alfa Aesar™ H50494MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50494MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 40848-53-1
    Molecular Formula C15H11ClN2
    Formula Weight 254.72
    MDL Number MFCD00172387
    Chemical Name or Material 1-Benzyl-4-chlorophthalazine

    Specifications

    Formula Weight 254.72
    Molecular Formula C15H11ClN2
    MDL Number MFCD00172387
    Quantity 250mg
    CAS 40848-53-1
    Chemical Name or Material 1-Benzyl-4-chlorophthalazine
    Assay 99%

    Description

    Applications
    1-Benzyl-4-chlorophthalazine is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit