Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyl-4-chlorophthalazine, 99%

Manufacturer:  Alfa Aesar™ H5049403

Quantity 1g

 View more versions of this product

Catalog No. AAH5049403

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
1-Benzyl-4-chlorophthalazine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

40848-53-1
254.72
1-Benzyl-4-chlorophthalazine
C15H11ClN2
MFCD00172387

Specifications

40848-53-1
C15H11ClN2
MFCD00172387
99%
1g
254.72
1-Benzyl-4-chlorophthalazine
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit