Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-cyano-4-hydroxypiperidine 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
6094-60-6
C13H16N2O
216.28
XQRLXUYZKZXSBN-UHFFFAOYSA-N
1-benzyl-4-cyano-4-hydroxypiperidine,maybridge1_006722,acmc-20aoxx,1-benzyl-4-hydroxyisonipecotonitrile,4-hydroxy-1-benzylpiperidine-4-carbonitrile,1-benyl-4-hydroxy-piperidine-4-carbonitrile,1-benzyl-4-hydroxy-4-piperidinecarbonitrile,1-benzyl-4-hydroxy-piperidine-4-carbonitrile,4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
80189
1-benzyl-4-hydroxypiperidine-4-carbonitrile
C1CN(CCC1(C#N)O)CC2=CC=CC=C2
6094-60-6
C13H16N2O
216.28
XQRLXUYZKZXSBN-UHFFFAOYSA-N
1-benzyl-4-cyano-4-hydroxypiperidine,maybridge1_006722,acmc-20aoxx,1-benzyl-4-hydroxyisonipecotonitrile,4-hydroxy-1-benzylpiperidine-4-carbonitrile,1-benyl-4-hydroxy-piperidine-4-carbonitrile,1-benzyl-4-hydroxy-4-piperidinecarbonitrile,1-benzyl-4-hydroxy-piperidine-4-carbonitrile,4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
80189
1-benzyl-4-hydroxypiperidine-4-carbonitrile
C1CN(CCC1(C#N)O)CC2=CC=CC=C2
Infrared Spectrum Authentic
Melting Point 95°C to 102°C
Packaging Glass bottle
Quantity 10g
Color Yellow to Beige
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 216.28
Physical Form Crystalline Powder and Chunks
Percent Purity ≥97.5%
Chemical Name or Material 1-Benzyl-4-cyano-4-hydroxypiperidine, 98%
Show More Show Less

EINECSNumber : 228-043-2

TSCA : TSCA