Alfa Aesar™ 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride, 99%

Manufacturer: Alfa Aesar™ H2741106

Quantity 5g

 View more versions of this product

Catalog No. AAH2741106

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 56243-25-5
    Molecular Formula C19H20N2·HCl
    Formula Weight 312.85
    MDL Number MFCD00035361
    Synonym 1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
    Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride

    Specifications

    Formula Weight 312.85
    MDL Number MFCD00035361
    CAS 56243-25-5
    Melting Point 261° to 264°C
    Quantity 5g
    Molecular Formula C19H20N2·HCl
    Synonym 1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
    Assay 99%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit