Alfa Aesar™ 1-Benzyl-7-azaindole-3-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5194903

Quantity 1g

 View more versions of this product

Catalog No. AAH5194903

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C20H23BN2O2
    Formula Weight 334.22
    MDL Number MFCD16294550
    Synonym 1-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
    Chemical Name or Material 1-Benzyl-7-azaindole-3-boronic acid pinacol ester

    Specifications

    Formula Weight 334.22
    Molecular Formula C20H23BN2O2
    MDL Number MFCD16294550
    Synonym 1-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
    Quantity 1g
    Chemical Name or Material 1-Benzyl-7-azaindole-3-boronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit