Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzylindole-3-carboxylic acid, 95%

Click to view available options
Quantity:
1g
5g
27018-76-4
C16H13NO2
251.285
MFCD00057094
LVYDDRHDOKXFMW-UHFFFAOYSA-N
1-benzyl-1h-indole-3-carboxylic acid,indole-3-carboxylic acid, 1-benzyl,1h-indole-3-carboxylic acid, 1-phenylmethyl,pubchem7313,cbmicro_033327,cambridge id 5791025,5-22-03-00037 beilstein handbook reference,1-benzylindol-3-carboxylic acid,n-benzylindole-3-carboxylic acid,1h-indole-3-carboxylicacid, 1-phenylmethyl
33671
1-benzylindole-3-carboxylic acid
C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O
27018-76-4
C16H13NO2
251.285
MFCD00057094
LVYDDRHDOKXFMW-UHFFFAOYSA-N
1-benzyl-1h-indole-3-carboxylic acid,indole-3-carboxylic acid, 1-benzyl,1h-indole-3-carboxylic acid, 1-phenylmethyl,pubchem7313,cbmicro_033327,cambridge id 5791025,5-22-03-00037 beilstein handbook reference,1-benzylindol-3-carboxylic acid,n-benzylindole-3-carboxylic acid,1h-indole-3-carboxylicacid, 1-phenylmethyl
33671
1-benzylindole-3-carboxylic acid
C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O
Quantity 5g
Beilstein 404957
Formula Weight 251.29
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 1-Benzylindole-3-carboxylic acid

RTECSNumber : NL5995800

storageNote1 : -30° to -10°C