Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylindole-3-carboxylic acid, 98%, Thermo Scientific™

Catalog Number ac428260010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
27018-76-4
C16H13NO2
251.28
MFCD00057094
LVYDDRHDOKXFMW-UHFFFAOYSA-N
1-benzyl-1h-indole-3-carboxylic acid, indole-3-carboxylic acid, 1-benzyl, 1h-indole-3-carboxylic acid, 1-phenylmethyl, pubchem7313, cbmicro_033327, cambridge id 5791025, 5-22-03-00037 beilstein handbook reference, 1-benzylindol-3-carboxylic acid, n-benzylindole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylmethyl
33671
1-benzylindole-3-carboxylic acid
C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O
27018-76-4
C16H13NO2
251.28
MFCD00057094
LVYDDRHDOKXFMW-UHFFFAOYSA-N
1-benzyl-1h-indole-3-carboxylic acid, indole-3-carboxylic acid, 1-benzyl, 1h-indole-3-carboxylic acid, 1-phenylmethyl, pubchem7313, cbmicro_033327, cambridge id 5791025, 5-22-03-00037 beilstein handbook reference, 1-benzylindol-3-carboxylic acid, n-benzylindole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylmethyl
33671
1-benzylindole-3-carboxylic acid
C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 251.28
Percent Purity 98%
Chemical Name or Material 1-Benzylindole-3-carboxylic acid

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit