Alfa Aesar™ 1-Benzyloxy-2-methyl-3-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ H5589903

Quantity 1g

 View more versions of this product

Catalog No. AAH5589903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20876-37-3
    Formula Weight 243.26
    MDL Number MFCD00100652
    Chemical Name or Material 1-Benzyloxy-2-methyl-3-nitrobenzene

    Specifications

    Formula Weight 243.26
    Melting Point 60° to 63°C
    Physical Form Powder
    MDL Number MFCD00100652
    CAS 20876-37-3
    Color Yellow to pale brown
    Quantity 1g
    Chemical Name or Material 1-Benzyloxy-2-methyl-3-nitrobenzene
    Assay 98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit