Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxy-4-bromobenzene, 97%

Click to view available options
Quantity:
1g
5g
25g
6793-92-6
C13H11BrO
263.134
MFCD00028016
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene,1-benzyloxy-4-bromobenzene,1-bromo-4-phenylmethoxy benzene,1-bromo-4-benzyloxybenzene,4-benzyloxy bromobenzene,1-benzyloxy-4-bromo-benzene,1-bromo-4-phenylmethyl oxy benzene,4-benzyloxy-bromobenzene,p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br
6793-92-6
C13H11BrO
263.134
MFCD00028016
OUQSGILAXUXMGI-UHFFFAOYSA-N
4-benzyloxybromobenzene,1-benzyloxy-4-bromobenzene,1-bromo-4-phenylmethoxy benzene,1-bromo-4-benzyloxybenzene,4-benzyloxy bromobenzene,1-benzyloxy-4-bromo-benzene,1-bromo-4-phenylmethyl oxy benzene,4-benzyloxy-bromobenzene,p-benzyloxy bromobenzene
138835
1-bromo-4-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)Br
Melting Point 61°C to 63°C
Quantity 1g
Boiling Point 166°C to 167°C (3mmHg)
Beilstein 1371040
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 1-Benzyloxy-4-bromobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning