Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 98%

Click to view available options
Quantity:
250mg
1g
286961-15-7
C19H26BNO4
343.23
MFCD11521562
QDSFHRPYZPQWEJ-UHFFFAOYSA-N
benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester,n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester,benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate,benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate,n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,benzyl 4-4,4,5,5-tetrameth
11290836
benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3
286961-15-7
C19H26BNO4
343.23
MFCD11521562
QDSFHRPYZPQWEJ-UHFFFAOYSA-N
benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate,4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester,n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester,benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate,benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate,n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,benzyl 4-4,4,5,5-tetrameth
11290836
benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3
Color White
Quantity 1g
Melting Point 88°C to 90°C
Beilstein 8571588
Formula Weight 343.23
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C