Alfa Aesar™ 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine, 97%

Manufacturer: Alfa Aesar™ H27583MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27583MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C13H13FN2O2
    Formula Weight 248.25
    MDL Number MFCD08458685
    UN Number UN3439
    Synonym Benzyl (2S,4R)-2-cyano-4-fluoro-pyrrolidine-1-carboxylate; N-Cbz-trans-4-fluoro-L-prolinonitrile
    Chemical Name or Material 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine

    Specifications

    Formula Weight 248.25
    Molecular Formula C13H13FN2O2
    UN Number UN3439
    Melting Point 68° to 72°C
    Health Hazard 1 Danger
    Quantity 250mg
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    MDL Number MFCD08458685
    Synonym Benzyl (2S,4R)-2-cyano-4-fluoro-pyrrolidine-1-carboxylate; N-Cbz-trans-4-fluoro-L-prolinonitrile
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 1-Benzyloxycarbonyl-(2S,4R)-2-cyano-4-fluoropyrrolidine
    Assay 97%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit