Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonyl-3-bromopyrrolidine, tech. 90%

Click to view available options
Quantity:
250mg
1g
5g
220212-12-4
C12H14BrNO2
284.153
MFCD13181593
LWEKWWHCQBWMQC-UHFFFAOYSA-N
1-cbz-3-bromopyrrolidine,3-bromo-pyrrolidine-1-carboxylic acid benzyl ester,3-bromopyrrolidine-1-carboxylic acid benzyl ester,cbz-3-bromopyrrolidine
53487977
benzyl 3-bromopyrrolidine-1-carboxylate
C1CN(CC1Br)C(=O)OCC2=CC=CC=C2
220212-12-4
C12H14BrNO2
284.153
MFCD13181593
LWEKWWHCQBWMQC-UHFFFAOYSA-N
1-cbz-3-bromopyrrolidine,3-bromo-pyrrolidine-1-carboxylic acid benzyl ester,3-bromopyrrolidine-1-carboxylic acid benzyl ester,cbz-3-bromopyrrolidine
53487977
benzyl 3-bromopyrrolidine-1-carboxylate
C1CN(CC1Br)C(=O)OCC2=CC=CC=C2
Applications
A commonly used as an organic reagent and intermediate for pharmaceutical.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Refractive Index 1.5570
Quantity 250mg
Density 1.395g/mL
Color Brown to Yellow
Formula Weight 284.15
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Benzyloxycarbonyl-3-bromopyrrolidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C