Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonyl-3-bromopyrrolidine, tech. 90%

Click to view available options
Quantity:
250mg
1g
5g
220212-12-4
C12H14BrNO2
284.153
MFCD13181593
LWEKWWHCQBWMQC-UHFFFAOYSA-N
1-cbz-3-bromopyrrolidine,3-bromo-pyrrolidine-1-carboxylic acid benzyl ester,3-bromopyrrolidine-1-carboxylic acid benzyl ester,cbz-3-bromopyrrolidine
53487977
benzyl 3-bromopyrrolidine-1-carboxylate
C1CN(CC1Br)C(=O)OCC2=CC=CC=C2
220212-12-4
C12H14BrNO2
284.153
MFCD13181593
LWEKWWHCQBWMQC-UHFFFAOYSA-N
1-cbz-3-bromopyrrolidine,3-bromo-pyrrolidine-1-carboxylic acid benzyl ester,3-bromopyrrolidine-1-carboxylic acid benzyl ester,cbz-3-bromopyrrolidine
53487977
benzyl 3-bromopyrrolidine-1-carboxylate
C1CN(CC1Br)C(=O)OCC2=CC=CC=C2
Applications
A commonly used as an organic reagent and intermediate for pharmaceutical.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Flash Point 110°C (230°F)
Density 1.395 g/mL
Refractive Index 1.557
Quantity 5g
Color Brown to Yellow
Formula Weight 284.15
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Benzyloxycarbonyl-3-bromopyrrolidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen