Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonyl-4-oxo-L-proline, 97%

Click to view available options
Quantity:
250mg
1g
5g
64187-47-9
C13H13NO5
263.249
MFCD07367987
RPLLCMZOIFOBIF-NSHDSACASA-N
n-carbobenzyloxy-4-keto-l-proline,n-carbobenzoxy-4-oxo-l-proline,s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,1-cbz-4-oxo-l-proline,s-1-z-4-oxopyrrolidine-2-carboxylic acid,s-1-benzyloxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,2s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,2s-4-oxo-1,2-pyrrolidinedicarboxylic acid 1-phenylmethyl ester,n-cbz-4-ketoproline,pubchem14286
11076282
(2S)-4-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1C(N(CC1=O)C(=O)OCC2=CC=CC=C2)C(=O)O
64187-47-9
C13H13NO5
263.249
MFCD07367987
RPLLCMZOIFOBIF-NSHDSACASA-N
n-carbobenzyloxy-4-keto-l-proline,n-carbobenzoxy-4-oxo-l-proline,s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,1-cbz-4-oxo-l-proline,s-1-z-4-oxopyrrolidine-2-carboxylic acid,s-1-benzyloxycarbonyl-4-oxopyrrolidine-2-carboxylic acid,2s-1-benzyloxy carbonyl-4-oxopyrrolidine-2-carboxylic acid,2s-4-oxo-1,2-pyrrolidinedicarboxylic acid 1-phenylmethyl ester,n-cbz-4-ketoproline,pubchem14286
11076282
(2S)-4-oxo-1-phenylmethoxycarbonylpyrrolidine-2-carboxylic acid
C1C(N(CC1=O)C(=O)OCC2=CC=CC=C2)C(=O)O
Formula Weight 263.24
Physical Form Powder
Percent Purity 97%
Color Brown to White
Melting Point 95°C to 100°C
Quantity 5g
Chemical Name or Material 1-Benzyloxycarbonyl-4-oxo-L-proline