Alfa Aesar™ 1-(Benzyloxycarbonyl)homopiperazine, 96%

Manufacturer: Alfa Aesar™ H55024AC

Quantity 25mL

 View more versions of this product

Catalog No. AAH55024AC

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 117009-97-9
    Molecular Formula C13H18N2O2
    Formula Weight 234.3
    MDL Number MFCD01863703
    Synonym Benzyl homopiperazine-1-carboxylate
    Chemical Name or Material 1-(Benzyloxycarbonyl)homopiperazine

    Specifications

    Formula Weight 234.3
    Molecular Formula C13H18N2O2
    MDL Number MFCD01863703
    Flash Point 110°C (230°F)
    CAS 117009-97-9
    Synonym Benzyl homopiperazine-1-carboxylate
    Boiling Point 130°C (0.5mmHg)
    Quantity 25mL
    Chemical Name or Material 1-(Benzyloxycarbonyl)homopiperazine
    Assay 96%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit