Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonyl-L-prolinamide, 95%

Click to view available options
Quantity:
5g
25g
100g
34079-31-7
C13H16N2O3
248.282
MFCD00020830
ZCGHEBMEQXMRQL-NSHDSACASA-N
z-pro-nh2,cbz-l-prolinamide,s-1-n-cbz-prolinamide,s-benzyl 2-carbamoylpyrrolidine-1-carboxylate,cbz-pro-nh2,z-l-prolinamide,z-l-proline amide,carbobenzyloxy-l-prolinamide,benzyl 2s-2-carbamoylpyrrolidine-1-carboxylate,benzyloxycarbonyl-l-proline amide
6950961
benzyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)N
34079-31-7
C13H16N2O3
248.282
MFCD00020830
ZCGHEBMEQXMRQL-NSHDSACASA-N
z-pro-nh2,cbz-l-prolinamide,s-1-n-cbz-prolinamide,s-benzyl 2-carbamoylpyrrolidine-1-carboxylate,cbz-pro-nh2,z-l-prolinamide,z-l-proline amide,carbobenzyloxy-l-prolinamide,benzyl 2s-2-carbamoylpyrrolidine-1-carboxylate,benzyloxycarbonyl-l-proline amide
6950961
benzyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
C1CC(N(C1)C(=O)OCC2=CC=CC=C2)C(=O)N
Melting Point 90°C to 95°C
Quantity 5g
Beilstein 88577
Formula Weight 248.28
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 1-Benzyloxycarbonyl-L-prolinamide

storageNote1 : 2° to 8°C