Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
188527-21-1
C13H15NO4
249.266
MFCD02179036
JSASVUTVTRNJHA-UHFFFAOYSA-N
1-n-cbz-pyrrolidine-3-carboxylic acid,n-cbz-pyrrolidine-3-carboxylic acid,1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid,1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid,1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid,1-cbz-pyrrolidine-3-carboxylic acid,1-n-cbz-beta-proline,1-n-cbz-belta-proline,1-cbz-3-pyrrolidinecarboxylic acid,3-carboxy-1-cbz-pyrrolidine
18538483
1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid
C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2
188527-21-1
C13H15NO4
249.266
MFCD02179036
JSASVUTVTRNJHA-UHFFFAOYSA-N
1-n-cbz-pyrrolidine-3-carboxylic acid,n-cbz-pyrrolidine-3-carboxylic acid,1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid,1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid,1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid,1-cbz-pyrrolidine-3-carboxylic acid,1-n-cbz-beta-proline,1-n-cbz-belta-proline,1-cbz-3-pyrrolidinecarboxylic acid,3-carboxy-1-cbz-pyrrolidine
18538483
1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid
C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2
Quantity 250mg
Beilstein 13268300
Formula Weight 249.27
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning