Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Benzyloxycarbonylmethyl-N-Boc-D-histidine, 98%

Click to view available options
Quantity:
1g
5g
25g
99310-01-7
C19H25N3O5
375.425
MFCD00076940
LPVKZCHCZSFTOJ-MRXNPFEDSA-N
boc-d-his bom-oh,boc-d-his 3-bom-oh,boc-d-his-bom-oh,n,a-boc-,eth-bom-d-histidine,boc-np-benzyloxymethyl-d-histidine,n,a-boc-n im-benzyloxymethyl-d-histidine,boc-d-his bom-oh tlc,2r-3-3-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
7018759
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC2=CC=CC=C2)C(=O)O
99310-01-7
C19H25N3O5
375.425
MFCD00076940
LPVKZCHCZSFTOJ-MRXNPFEDSA-N
boc-d-his bom-oh,boc-d-his 3-bom-oh,boc-d-his-bom-oh,n,a-boc-,eth-bom-d-histidine,boc-np-benzyloxymethyl-d-histidine,n,a-boc-n im-benzyloxymethyl-d-histidine,boc-d-his bom-oh tlc,2r-3-3-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid
7018759
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[3-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CN1COCC2=CC=CC=C2)C(=O)O
Formula Weight 375.43
Physical Form Powder
Percent Purity 98%
Color White
Quantity 25g
Chemical Name or Material 1-Benzyloxycarbonylmethyl-N-Boc-D-histidine