Alfa Aesar™ 1-Boc-2-(aminoethyl)pyrrolidine, 97%

Manufacturer: Alfa Aesar™ H28148MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28148MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 370069-29-7
    Molecular Formula C11H22N2O2
    Formula Weight 214.31
    MDL Number MFCD04115289
    UN Number UN2735
    Synonym 1-tert-Butoxycarbonyl-2-(2-aminoethyl)pyrrolidine; tert-Butyl 2-(2-aminoethyl)pyrrolidine-1-carboxylate
    Chemical Name or Material 1-Boc-2-(aminoethyl)pyrrolidine

    Specifications

    Formula Weight 214.31
    Molecular Formula C11H22N2O2
    MDL Number MFCD04115289
    Quantity 250mg
    CAS 370069-29-7
    UN Number UN2735
    Synonym 1-tert-Butoxycarbonyl-2-(2-aminoethyl)pyrrolidine; tert-Butyl 2-(2-aminoethyl)pyrrolidine-1-carboxylate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material 1-Boc-2-(aminoethyl)pyrrolidine
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit