Alfa Aesar™ (+/-)-1-Boc-2-(aminomethyl)pyrrolidine, tech. 85%

Manufacturer: Alfa Aesar™ H28650MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28650MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 177911-87-4
    Molecular Formula C10H20N2O2
    Formula Weight 200.28
    MDL Number MFCD01317793
    UN Number UN2735
    Synonym (+/-)-1-tert-Butoxycarbonyl-2-(aminomethyl)pyrrolidine; tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
    Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine
    Grade Technical Grade

    Specifications

    Formula Weight 200.28
    Molecular Formula C10H20N2O2
    MDL Number MFCD01317793
    Quantity 250mg
    UN Number UN2735
    Synonym (+/-)-1-tert-Butoxycarbonyl-2-(aminomethyl)pyrrolidine; tert-Butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 177911-87-4
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine
    Assay 85%
    Grade Technical Grade

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit