Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Boc-2-cyanopiperidine, 96%

Click to view available options
Quantity:
1g
5g
153749-89-4
C11H18N2O2
210.277
MFCD01861221
LKAJZBMOVZIKHA-UHFFFAOYSA-N
n-boc-2-cyanopiperidine,1-boc-2-cyanopiperidine,1-n-boc-2-cyanopiperidine,+/--1-n-boc-2-cyano-piperidine,1-n-boc-2-cyano-piperidine,2-cyano-piperidine-1-carboxylic acid tert-butyl ester,n-boc-2-piperidinecarbonitrile,1-boc-piperidine-2-carbonitrile,1-piperidinecarboxylic acid, 2-cyano-, 1,1-dimethylethyl ester,tert-butyl 2-cyanopiperidinecarboxylate
2756784
tert-butyl 2-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1C#N
153749-89-4
C11H18N2O2
210.277
MFCD01861221
LKAJZBMOVZIKHA-UHFFFAOYSA-N
n-boc-2-cyanopiperidine,1-boc-2-cyanopiperidine,1-n-boc-2-cyanopiperidine,+/--1-n-boc-2-cyano-piperidine,1-n-boc-2-cyano-piperidine,2-cyano-piperidine-1-carboxylic acid tert-butyl ester,n-boc-2-piperidinecarbonitrile,1-boc-piperidine-2-carbonitrile,1-piperidinecarboxylic acid, 2-cyano-, 1,1-dimethylethyl ester,tert-butyl 2-cyanopiperidinecarboxylate
2756784
tert-butyl 2-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1C#N
Melting Point 62°C to 67°C
Quantity 1g
Color Brown to White
UN Number UN3439
Formula Weight 210.27
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 1-Boc-2-cyanopiperidine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : 2° to 8°C