Alfa Aesar™ 1-Boc-3,3-difluoropyrrolidine, 98%

Manufacturer: Alfa Aesar™ H2742203

Quantity 1g

 View more versions of this product

Catalog No. AAH2742203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 195447-25-7
    Molecular Formula C9H15F2NO2
    Formula Weight 207.22
    MDL Number MFCD06797010
    Synonym tert-Butyl 3,3-difluoropyrroline-1-carboxylate
    Chemical Name or Material 1-Boc-3,3-difluoropyrrolidine

    Specifications

    Formula Weight 207.22
    Molecular Formula C9H15F2NO2
    MDL Number MFCD06797010
    Quantity 1g
    Synonym tert-Butyl 3,3-difluoropyrroline-1-carboxylate
    CAS 195447-25-7
    Chemical Name or Material 1-Boc-3,3-difluoropyrrolidine
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit