Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-3-(aminomethyl)azetidine, 95%, Thermo Scientific™

Click to view available options
Quantity:
250mg
1g
325775-44-8
C9H18N2O2
186.25
XSJPKMUFBHSIRA-UHFFFAOYSA-N
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
10679125
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
CC(C)(C)OC(=O)N1CC(C1)CN
325775-44-8
C9H18N2O2
186.25
XSJPKMUFBHSIRA-UHFFFAOYSA-N
1-boc-3-aminomethyl azetidine, tert-butyl 3-aminomethyl azetidine-1-carboxylate, 1-boc-3-aminomethylazetidine, 3-aminomethyl-1-n-boc-azetidine, 1-n-boc-3-aminomethylazetidine, 3-aminomethyl-azetidine-1-carboxylic acid tert-butyl ester, 1-boc-3-aminomethyl-azetidine, 3-aminomethyl azetidine, n1-boc protected, tert-butyl 3-aminomethyl azetidinecarboxylate, 1-azetidinecarboxylic acid, 3-aminomethyl-, 1,1-dimethylethyl ester
10679125
tert-butyl 3-(aminomethyl)azetidine-1-carboxylate
CC(C)(C)OC(=O)N1CC(C1)CN
Quantity 1g
Packaging Glass Bottle
Flash Point >110°C
Assay Percent Range 94% min. (GC)
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material 1-BOC-3-(aminomethyl)azetidine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

RUO – Research Use Only