Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-3-Aminopiperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
144243-24-3
C10H20N2O2
200.28
AKQXKEBCONUWCL-UHFFFAOYSA-N
1-boc-3-aminopiperidine,n-boc-3-aminopiperidine,3-amino-1-n-boc-piperidine,+/--3-amino-1-n-boc-piperidine,3-amino-1-boc-piperidine,1-n-boc-3-aminopiperidine,3-amino-piperidine-1-carboxylic acid tert-butyl ester,1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester,3-amino-1-tert-butoxycarbonylpiperidine,3-aminopiperidine-1-carboxylic acid tert-butyl ester
545809
tert-butyl 3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
144243-24-3
C10H20N2O2
200.28
AKQXKEBCONUWCL-UHFFFAOYSA-N
1-boc-3-aminopiperidine,n-boc-3-aminopiperidine,3-amino-1-n-boc-piperidine,+/--3-amino-1-n-boc-piperidine,3-amino-1-boc-piperidine,1-n-boc-3-aminopiperidine,3-amino-piperidine-1-carboxylic acid tert-butyl ester,1-piperidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester,3-amino-1-tert-butoxycarbonylpiperidine,3-aminopiperidine-1-carboxylic acid tert-butyl ester
545809
tert-butyl 3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
Packaging Glass bottle
Color Colorless to Yellow
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 200.28
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-BOC-3-Aminopiperidine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning