Alfa Aesar™ 1-Boc-3-azetidinone, 95%

Manufacturer: Alfa Aesar™ H5082603

Quantity 1g

 View more versions of this product

Catalog No. AAH5082603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 398489-26-4
    Molecular Formula C8H13NO3
    Formula Weight 171.2
    MDL Number MFCD01861741
    Synonym tert-Butyl 3-oxoazetidine-1-carboxylate
    Chemical Name or Material 1-Boc-3-azetidinone

    Specifications

    Formula Weight 171.2
    Molecular Formula C8H13NO3
    Melting Point 49° to 52°C
    MDL Number MFCD01861741
    Quantity 1g
    CAS 398489-26-4
    Synonym tert-Butyl 3-oxoazetidine-1-carboxylate
    Chemical Name or Material 1-Boc-3-azetidinone
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit