Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Boc-3-cyanopiperidine, 96%

Click to view available options
Quantity:
1g
5g
91419-53-3
C11H18N2O2
210.277
MFCD01861222
UEFZTXGFHKPSFS-UHFFFAOYSA-N
1-n-boc-3-cyanopiperidine,n-boc-3-cyanopiperidine,1-boc-3-cyanopiperidine,+/--1-n-boc-3-cyano-piperidine,n-boc-3-cyano-piperidine,1-n-boc-3-cyano-piperidine,3-cyano-piperidine-1-carboxylic acid tert-butyl ester,1-n-boc-piperidine-3-carbonitrile,1-tert-butoxycarbonyl-3-cyanopiperidine,tert-butyl-3-cyanopiperidine-1-carboxylate
2756785
tert-butyl 3-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)C#N
91419-53-3
C11H18N2O2
210.277
MFCD01861222
UEFZTXGFHKPSFS-UHFFFAOYSA-N
1-n-boc-3-cyanopiperidine,n-boc-3-cyanopiperidine,1-boc-3-cyanopiperidine,+/--1-n-boc-3-cyano-piperidine,n-boc-3-cyano-piperidine,1-n-boc-3-cyano-piperidine,3-cyano-piperidine-1-carboxylic acid tert-butyl ester,1-n-boc-piperidine-3-carbonitrile,1-tert-butoxycarbonyl-3-cyanopiperidine,tert-butyl-3-cyanopiperidine-1-carboxylate
2756785
tert-butyl 3-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)C#N
Sensitivity Moisture Sensitive
Melting Point 60°C to 65°C
Color White to Yellow
Quantity 1g
UN Number UN3439
Formula Weight 210.27
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 1-Boc-3-cyanopiperidine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : 2° to 8°C