Alfa Aesar™ 1-Boc-3-cyanopyrrolidine, 99%

Manufacturer: Alfa Aesar™ H50082MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50082MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 476493-40-0
    Molecular Formula C10H16N2O2
    Formula Weight 196.25
    MDL Number MFCD03001709
    Synonym 1-Boc-pyrrolidine-3-carbonitrile; tert-Butyl 3-cyanopyrrolidine-1-carboxylate
    Chemical Name or Material 1-Boc-3-cyanopyrrolidine

    Specifications

    Formula Weight 196.25
    Molecular Formula C10H16N2O2
    MDL Number MFCD03001709
    Quantity 250mg
    Synonym 1-Boc-pyrrolidine-3-carbonitrile; tert-Butyl 3-cyanopyrrolidine-1-carboxylate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 476493-40-0
    Chemical Name or Material 1-Boc-3-cyanopyrrolidine
    Assay 99%

    GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit