Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-3-iodoazetidine, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
254454-54-1
C8H14INO2
283.1
XPDIKRMPZNLBAC-UHFFFAOYSA-N
1-boc-3-iodoazetidine,1-boc-3-iodo-azetidine,1-boc-3-iodo azetidine,n-boc-3-iodoazetidine,3-iodo-azetidine-1-carboxylic acid tert-butyl ester,3-iodoazetidine, n-boc protected,1-n-boc-3-iodoazetidine,1-tert-butoxycarbonyl-3-iodoazetidine,tert-butyl 3-iodo-1-azetidinecarboxylate,tert-butyl 3-iodoazetidinecarboxylate
11000522
tert-butyl 3-iodoazetidine-1-carboxylate
CC(C)(C)OC(=O)N1CC(C1)I
254454-54-1
C8H14INO2
283.1
XPDIKRMPZNLBAC-UHFFFAOYSA-N
1-boc-3-iodoazetidine,1-boc-3-iodo-azetidine,1-boc-3-iodo azetidine,n-boc-3-iodoazetidine,3-iodo-azetidine-1-carboxylic acid tert-butyl ester,3-iodoazetidine, n-boc protected,1-n-boc-3-iodoazetidine,1-tert-butoxycarbonyl-3-iodoazetidine,tert-butyl 3-iodo-1-azetidinecarboxylate,tert-butyl 3-iodoazetidinecarboxylate
11000522
tert-butyl 3-iodoazetidine-1-carboxylate
CC(C)(C)OC(=O)N1CC(C1)I
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 94% min. (GC)
Formula Weight 283.1
Percent Purity 95%
Chemical Name or Material 1-BOC-3-iodoazetidine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning