Alfa Aesar™ 1-Boc-3-iodoindole, 95%

Manufacturer: Alfa Aesar™ H27022MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27022MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 192189-07-4
    Molecular Formula C13H14INO2
    Formula Weight 343.16
    MDL Number MFCD05864781
    Synonym tert-Butyl 3-iodo-1H-indole-1-carboxylate
    Chemical Name or Material 1-Boc-3-iodoindole

    Specifications

    Formula Weight 343.16
    Molecular Formula C13H14INO2
    MDL Number MFCD05864781
    CAS 192189-07-4
    Synonym tert-Butyl 3-iodo-1H-indole-1-carboxylate
    Quantity 250mg
    Recommended Storage −30° to −10°C
    Chemical Name or Material 1-Boc-3-iodoindole
    Assay 95%

    Description

    Applications
    1-Boc-3-iodoindole is used as a pharmaceutical intermediate and also used in ceramic manufacturing industry.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light Sensitive.

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit