Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-3-methylaminopyrrolidine, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
454712-26-6
C10H20N2O2
200.28
OKUCEQDKBKYEJY-UHFFFAOYSA-N
1-boc-3-methylaminopyrrolidine, tert-butyl 3-methylamino pyrrolidine-1-carboxylate, 1-boc-3-methylamino-pyrrolidine, 1-boc-3-methylamino pyrrolidine, 3-methylamino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, s-1-boc-3-methylamino pyrrolidine, pubchem11215, acmc-1aqtj, acmc-20a85z
45089533
tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)NC
454712-26-6
C10H20N2O2
200.28
OKUCEQDKBKYEJY-UHFFFAOYSA-N
1-boc-3-methylaminopyrrolidine, tert-butyl 3-methylamino pyrrolidine-1-carboxylate, 1-boc-3-methylamino-pyrrolidine, 1-boc-3-methylamino pyrrolidine, 3-methylamino-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 3-methylamino-, 1,1-dimethylethyl ester, s-1-boc-3-methylamino pyrrolidine, pubchem11215, acmc-1aqtj, acmc-20a85z
45089533
tert-butyl 3-(methylamino)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)NC
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass Bottle
Assay Percent Range 96% min. (GC)
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material 1-BOC-3-methylaminopyrrolidine

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

RUO – Research Use Only