Alfa Aesar™ 1-Boc-4-(2-methoxycarbonylphenyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5530206

Quantity 5g

 View more versions of this product

Catalog No. AAH5530206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 870703-74-5
    Formula Weight 320.39
    MDL Number MFCD07369760
    UN Number UN2810
    Synonym Methyl 2-(Boc-1-piperazinyl)benzoate
    Chemical Name or Material 1-Boc-4-(2-methoxycarbonylphenyl)piperazine

    Specifications

    Formula Weight 320.39
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2810
    MDL Number MFCD07369760
    CAS 870703-74-5
    Color Yellow to orange
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Refractive Index 1.5320
    Physical Form Liquid
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Synonym Methyl 2-(Boc-1-piperazinyl)benzoate
    Chemical Name or Material 1-Boc-4-(2-methoxycarbonylphenyl)piperazine
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit