Alfa Aesar™ 1-Boc-4-(3-hydroxypropyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5554703

Quantity 1g

 View more versions of this product

Catalog No. AAH5554703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 132710-90-8
    Formula Weight 244.33
    MDL Number MFCD06798090
    Chemical Name or Material 1-Boc-4-(3-hydroxypropyl)piperazine

    Specifications

    Melting Point 73° to 77°C
    Formula Weight 244.33
    MDL Number MFCD06798090
    Quantity 1g
    Physical Form Solid
    CAS 132710-90-8
    Chemical Name or Material 1-Boc-4-(3-hydroxypropyl)piperazine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit