Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Boc-4-(4-aminophenyl)piperazine, 97%

Click to view available options
Quantity:
250mg
1g
5g
170911-92-9
C15H23N3O2
277.368
MFCD04115065
RXFHRKPNLPBDGE-UHFFFAOYSA-N
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate,4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester,4-4-boc-piperazin-1-yl aniline,1-boc-4-4-aminophenyl piperazine,1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester,4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester,1-boc-4-4'-aminophenyl piperazine dihydrochloride,1-boc-4-4-aminophenyl-piperazine,pubchem12198
11011301
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N
170911-92-9
C15H23N3O2
277.368
MFCD04115065
RXFHRKPNLPBDGE-UHFFFAOYSA-N
tert-butyl 4-4-aminophenyl piperazine-1-carboxylate,4-4-aminophenyl piperazine-1-carboxylic acid tert-butyl ester,4-4-boc-piperazin-1-yl aniline,1-boc-4-4-aminophenyl piperazine,1-piperazinecarboxylic acid, 4-4-aminophenyl-, 1,1-dimethylethyl ester,4-4-aminophenyl-piperazine-1-carboxylic acid tert-butyl ester,1-boc-4-4'-aminophenyl piperazine dihydrochloride,1-boc-4-4-aminophenyl-piperazine,pubchem12198
11011301
tert-butyl 4-(4-aminophenyl)piperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)N
Applications
1-Boc-4-(4-aminophenyl)piperazine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Solubility Information Slightly soluble in water.
Formula Weight 277.36
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-Boc-4-(4-aminophenyl)piperazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures