Alfa Aesar™ 1-Boc-4-(4-formylphenyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5541306

Quantity 5g

 View more versions of this product

Catalog No. AAH5541306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 197638-83-8
    Molecular Formula C16H22N2O3
    Formula Weight 290.36
    MDL Number MFCD05864663
    UN Number UN2811
    Synonym 4-(1-Boc-1-piperazinyl)benzaldehyde
    Chemical Name or Material 1-Boc-4-(4-formylphenyl)piperazine

    Specifications

    Formula Weight 290.36
    Quantity 5g
    Molecular Formula C16H22N2O3
    Melting Point 118° to 121°C
    MDL Number MFCD05864663
    CAS 197638-83-8
    UN Number UN2811
    Synonym 4-(1-Boc-1-piperazinyl)benzaldehyde
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 1-Boc-4-(4-formylphenyl)piperazine
    Assay 97%

    GHS H Statement
    H301-H317
    Toxic if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit