Alfa Aesar™ 1-Boc-4-cyanopiperidine, 96%

Manufacturer: Alfa Aesar™ H2758006

Quantity 5g

 View more versions of this product

Catalog No. AAH2758006

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 91419-52-2
    Molecular Formula C11H18N2O2
    Formula Weight 210.27
    MDL Number MFCD01861223
    UN Number UN3439
    Synonym 1-Boc-piperidine-4-carbonitrile; tert-Butyl 4-cyanopiperidine-1-carboxylate
    Chemical Name or Material 1-Boc-4-cyanopiperidine

    Specifications

    Formula Weight 210.27
    Molecular Formula C11H18N2O2
    Melting Point 45° to 48°C
    MDL Number MFCD01861223
    Quantity 5g
    CAS 91419-52-2
    UN Number UN3439
    Synonym 1-Boc-piperidine-4-carbonitrile; tert-Butyl 4-cyanopiperidine-1-carboxylate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 1-Boc-4-cyanopiperidine
    Assay 96%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit