Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(Boc-amino)cyclobutanecarboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
120728-10-1
C10H17NO4
215.249
MFCD02682623
ROVVUKFHORPDSM-UHFFFAOYSA-N
n-boc-1-aminocyclobutanecarboxylic acid,1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid,1-boc-amino cyclobutanecarboxylic acid,boc-cyclovaline,boc-acbc-oh,1-n-boc-amino-cyclobutane carboxylic acid,1-tert-butoxycarbonylamino cyclobutanecarboxylic acid,boc-1-aminocyclobutane-1-carboxylic acid,n-boc-1-aminocyclobutane carboxylic acid,boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
120728-10-1
C10H17NO4
215.249
MFCD02682623
ROVVUKFHORPDSM-UHFFFAOYSA-N
n-boc-1-aminocyclobutanecarboxylic acid,1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid,1-boc-amino cyclobutanecarboxylic acid,boc-cyclovaline,boc-acbc-oh,1-n-boc-amino-cyclobutane carboxylic acid,1-tert-butoxycarbonylamino cyclobutanecarboxylic acid,boc-1-aminocyclobutane-1-carboxylic acid,n-boc-1-aminocyclobutane carboxylic acid,boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
Quantity 1g
Melting Point 129°C to 133°C
Beilstein 7994003
Formula Weight 215.24
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(Boc-amino)cyclobutanecarboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures