Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-Azetidine-3-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
142253-55-2
C9H15NO4
201.22
NCADHSLPNSTDMJ-UHFFFAOYSA-N
1-boc-azetidine-3-carboxylic acid,1-n-boc-3-azetidinecarboxylic acid,n-boc-azetidine-3-carboxylic acid,boc-azetidine-3-carboxylic acid,1-tert-butoxycarbonyl azetidine-3-carboxylic acid,1-tert-butoxy carbonyl azetidine-3-carboxylic acid,1-n-boc-azetidine-3-carboxylic acid,boc-aze 3-oh,azetidine-1,3-dicarboxylic acid mono-tert-butyl ester,n-boc-3-azetidine carboxylic acid
2755981
1-[(2-methylpropan-2-yl)oxycarbonyl]azetidine-3-carboxylic acid
CC(C)(C)OC(=O)N1CC(C1)C(=O)O
142253-55-2
C9H15NO4
201.22
NCADHSLPNSTDMJ-UHFFFAOYSA-N
1-boc-azetidine-3-carboxylic acid,1-n-boc-3-azetidinecarboxylic acid,n-boc-azetidine-3-carboxylic acid,boc-azetidine-3-carboxylic acid,1-tert-butoxycarbonyl azetidine-3-carboxylic acid,1-tert-butoxy carbonyl azetidine-3-carboxylic acid,1-n-boc-azetidine-3-carboxylic acid,boc-aze 3-oh,azetidine-1,3-dicarboxylic acid mono-tert-butyl ester,n-boc-3-azetidine carboxylic acid
2755981
1-[(2-methylpropan-2-yl)oxycarbonyl]azetidine-3-carboxylic acid
CC(C)(C)OC(=O)N1CC(C1)C(=O)O
Melting Point 105°C to 107°C
Quantity 500mg
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 201.22
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-BOC-Azetidine-3-carboxylic acid
Show More Show Less