Alfa Aesar™ 1-Boc-isonipecotic acid ethyl ester, 97+%

Manufacturer: Alfa Aesar™ B2539314

Quantity 25g

 View more versions of this product

Catalog No. AAB2539314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142851-03-4
    Molecular Formula C13H23NO4
    Formula Weight 257.33
    MDL Number MFCD01763998
    Synonym Ethyl N-Boc-piperidine-4-carboxylate; Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester
    Chemical Name or Material 1-Boc-isonipecotic acid ethyl ester

    Specifications

    Formula Weight 257.33
    MDL Number MFCD01763998
    Quantity 25g
    Flash Point >110°C (230°F)
    Molecular Formula C13H23NO4
    CAS 142851-03-4
    Synonym Ethyl N-Boc-piperidine-4-carboxylate; Piperidine-1,4-dicarboxylic acid 1-tert-butyl ester 4-ethyl ester
    Boiling Point 120° to 135°C (0.5mmHg)
    Recommended Storage 2° to 8°C
    Chemical Name or Material 1-Boc-isonipecotic acid ethyl ester
    Assay ≥97%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit