Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-Piperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
75844-69-8
C10H19NO2
185.27
RQCNHUCCQJMSRG-UHFFFAOYSA-N
1-boc-piperidine,n-boc-piperidine,1-tert-butoxycarbonyl piperidine,1-piperidinecarboxylic acid, 1,1-dimethylethyl ester,piperidine-1-carboxylic acid tert-butyl ester,tert-butyl piperidinecarboxylate,boc-piperidine,n-boc piperidine,pubchem7258,n-boc tetrahydropyridine
7010304
tert-butyl piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1
75844-69-8
C10H19NO2
185.27
RQCNHUCCQJMSRG-UHFFFAOYSA-N
1-boc-piperidine,n-boc-piperidine,1-tert-butoxycarbonyl piperidine,1-piperidinecarboxylic acid, 1,1-dimethylethyl ester,piperidine-1-carboxylic acid tert-butyl ester,tert-butyl piperidinecarboxylate,boc-piperidine,n-boc piperidine,pubchem7258,n-boc tetrahydropyridine
7010304
tert-butyl piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCCC1
Boiling Point 58°C (0.3 mbar)
Flash Point 86°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless to Yellow
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 185.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-BOC-Piperidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning